missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10402504
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10402504 25 g 25g
10077632 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10402504 Supplier Thermo Scientific Maybridge Supplier No. TL00486EB
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 4426-79-3
Molecular Formula C5H9NS
Molecular Weight (g/mol) 115.19
MDL Number MFCD00041108
InChI Key TUFJIDJGIQOYFY-UHFFFAOYNA-N
Synonym sec-butyl isothiocyanate, 2-butyl isothiocyanate, butane, 2-isothiocyanato, isothiocyanic acid sec-butyl ester, isothiocyanic acid, sec-butyl ester, butan-2-isothiocyanate, 2-butylisothiocyanate, s-butyl isothiocyanate, 2-isothiocyanato-butane
PubChem CID 78151
IUPAC Name 2-isothiocyanatobutane
SMILES CCC(C)N=C=S

Specifications

Assay Percent Range 97%
Quantity 25 g
Formula Weight 115.2
Chemical Name or Material sec-Butyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.