missing translation for 'onlineSavingsMsg'
Learn More

sec-Butylmalonic Acid Diethyl Ester, TRC

Product Code. 30336584
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30336584 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336584 Supplier Toronto Research Chemicals Supplier No. TRCB6926101G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material sec-Butylmalonic Acid Diethyl Ester
CAS 83-27-2
Molecular Formula C11H20O4
Quantity 1 g
Synonym (1-Methylpropyl)propanedioic Acid Diethyl Ester, Diethyl sec-Butylmalonate, NSC 127865, NSC 30679, NSC 41157, sec-Butylmalonic Acid Diethyl Ester
InChI Formula InChI=1S/C11H20O4/c1-5-8(4)9(10(12)14-6-2)11(13)15-7-3/h8-9H,5-7H2,1-4H3
SMILES CCOC(=O)C(C(C)CC)C(=O)OCC
IUPAC Name diethyl 2-butan-2-ylpropanedioate
Molecular Weight (g/mol) 216.27
Formula Weight 216.1362
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.