missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Secador™ 5.0 Auto-Desiccator Cabinet, Electric Non-Desiccant

Product Code. 17885392
Change view
Click to view available options
No. of Shelves:
3
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Shelves Unit Size
17885392 3 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17885392 Supplier Bel-Art™ Supplier No. H420571003
  • ONLINE EXCLUSIVE PRICE
    2150.00 €
    1902.53 € / Pack of 1
    Save 247.47 €
    11% Off
 Restricted Item
Estimated Shipment: 12-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Desiccant-free cabinet provides dry storage for humidity-sensitive products such as laboratory samples, reagents, optical equipment, electronics, and 3D printing filament.

  • High-density foam gaskets to keep ambient humidity out and dry air inside
  • Large clear polycarbonate panels are durable and allow for easy viewing of internal contents
  • LCD shows the internal temperature and humidity level
  • Includes 3 adjustable and removable shelves
  • Electric models keeps internal humidity at 20-40%

Specifications

Autoclavable Not Autoclavable
Color White
Cabinet Style Front-loading
Material Polycarbonate
Product Type Desiccator
Door Style Hinged
No. of Shelves 3
Shelf Positions Adjustable
Dimensions (D x W x H) Interior 34 cm x 28 cm x 50cm
Width (English) 11.8 in.
Width (Metric) 30 cm
Height (English) 25 in.
Height (Metric) 63.5 cm
Depth (English) 13 in.
Depth (Metric) 34 cm
Voltage 100/120 V or 200/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.