missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ SecinH3

Product Code. 18034292
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18034292 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18034292 Supplier R&D Systems™ Supplier No. 2849/5
  • ONLINE EXCLUSIVE PRICE
    166.77 €
    149.10 € / 5mg
    Save 17.67 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Sec7-specific GEF inhibitor; selective for cytohesin family

SecinH3 is a Sec7-specific GEF inhibitor that displays selectivity for small GEFs of the cytohesin family (IC50 values are 2.4, 5.4, 5.4, 5.6, 5.6, 65 and > 100 μM for hCyh2, hCyh1, mCyh3, hCyh3, drosophila steppke, yGea2-S7 and hEFA6-S7 respectively). Inhibits insulin signaling via inhibition of insulin receptor substrate protein (IRS) phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[4-[5-(1,3-Benzodioxol-5-yl)-3-methoxy-1H-1,2,4-triazol-1-yl]phenyl]-2-(phenylthio)acetamide
CAS 853625-60-2
Quantity 5 mg
Product Line Tocris
Target ARF GTPase Inhibitors
Molecular Formula C24H20N4O4S
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.