Learn More
3M™ SecureFit™ Anti-Scratch, Anti-Fog Safety Spectacles
Product Code. 15312004 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Amber
Blue Mirror
Clear
Gray
Indoor/Outdoor
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint Unit Size
15312004 Indoor/Outdoor Each
15322004 Blue Mirror Each
15371994 Clear Each
15302004 Amber Each
15391994 Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15312004 Supplier 3M™ Supplier No. 7100078988
  • ONLINE EXCLUSIVE PRICE
    19.05 €
    9.78 € / Pack of 1
    Save 9.27 €
    48% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Reduces slippage from head movements. 3M™ SecureFit™ Anti-Scratch, Anti-Fog Safety Spectacles provide a secure and comfortable fit with the 3M Pressure Diffusion Temple Technology.
  • Available in a range of lens tints
  • Protection against UV radiation
  • Naturally adjusts to individuals' head sizes
  • Rimless lenses and fixed-length temple arm to enhance frame comfort
  • Soft, adjustable nose pads for even greater personalization of fit
  • Get added comfort over the ears with padded temple touchpoints
  • Temple arms in a bold colour for an infusion of style
TRUSTED_SUSTAINABILITY
Specifications
Lens Tint Indoor/Outdoor
Product Type Goggles
No. per Pack 1 Ea.
Lens Material Polycarbonate