Learn More
3M™ SecureFit™ 500 Safety Glasses
Product Code. 17354680 Shop All 3M Products
Change view
Click to view available options
Model:
SF501AF-BLK-EU
SF501ASP-BLK-EU
SF501SGAF-BLK-EU
SF501SGAF-BLK-FM-EU
SF501SGAF-GRN-EU
SF501SGAF-RED-EU
SF502SGAF-BLK-EU
SF503SGAF-BLK-EU
SF505SGAF-DGR-EU
SF507SGAF-BLK-EU
SF509AF-BLK
SF517ASP-GRN-EU
SF528SGAF-DGR-EU
SF530ASP-GRN-EU
SF550ASP-GRN-EU
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
Unit Size:
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Lens Coating Unit Size
17354680 SF550ASP-GRN-EU Scratch-resistant Each
17374670 SF501SGAF-BLK-EU Anti-fog, Scratch-resistant Each
17384670 SF501SGAF-RED-EU Anti-fog, Scratch-resistant Each
17304680 SF509AF-BLK Anti-fog, Scratch-resistant Each
17314680 SF501SGAF-BLK-FM-EU Anti-fog, Scratch-resistant Each
17334680 SF501SGAF-GRN-EU Anti-fog, Scratch-resistant Each
17344680 SF517ASP-GRN-EU Scratch-resistant Each
17364680 SF501ASP-BLK-EU Scratch-resistant Each
17374680 SF503SGAF-BLK-EU Anti-fog, Scratch-resistant Each
17384680 SF530ASP-GRN-EU Scratch-resistant Each
17394680 SF507SGAF-BLK-EU Anti-fog, Scratch-resistant Each
17304690 SF505SGAF-DGR-EU Anti-fog, Scratch-resistant Each
17314690 SF501AF-BLK-EU Anti-fog, Scratch-resistant Each
17374690 SF528SGAF-DGR-EU Anti-fog, Scratch-resistant Each
17394690 SF502SGAF-BLK-EU Anti-fog, Scratch-resistant Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 17354680 Supplier 3M™ Supplier No. 7100243978
  • ONLINE EXCLUSIVE PRICE
    22.30 €
    15.45 € / Pack of 1
    Save 6.85 €
    30% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
3M™ SecureFit™ Safety Glasses 500 Series features a stylized lens shape with temples that automatically self-adjust to help provide a secure and comfortable fit.
  • Sleek and compatible 3M™ SecureFit™ Safety Glasses range with adjustable ratchet temples to adjust the lens for facial features and eyeglass position, designed to help improve the positioning when worn with respirators
  • 3M's innovative SecureFit™ Pressure Diffusion Temple technology design features slotted ribs that provide comfortable pressure equalization. The temples flex and self-adjust to the size of wearer's head
  • 3M™ Scotchgard™ Anti-Fog Coating provides superior anti-fog and anti-scratch properties (available on selected models)
  • Protection against UV up to 400 nm
  • Wraparound design improves the security of fit and prevents slipping
  • Flat and flexible temples combine easily with earmuffs and provide solid side protection
  • Easy to clean
  • Foam gasket available to help limit eye exposure to debris
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance (Europe) EN 166:2001
Lens Tint IR 5.0 Gray
Product Type Protective Eyewear
Frame Color Green, Black
Model SF550ASP-GRN-EU
Frame Material Polycarbonate
UV Protection UV400