Learn More
3M™ SecureFit™ Prescription Insert for 200 and 400 Series, RX-SF400
Product Code. 30784776 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Product Code. Lens Tint
30784776 Clear
1 options
Product Code. 30784776 Supplier 3M™ Supplier No. 7010528331
  • ONLINE EXCLUSIVE PRICE
    42.00 €
    26.94 € / Pack of 1
    Save 15.06 €
    35% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Prescription lens inserts are designed to add prescription lenses to 3M™ safety glasses and goggles.
  • Compatible with 3M™ safety glasses and goggles.
  • Holds polycarbonate prescription lenses up to ±6 dioptres.
  • Maintains full eye protection while improving vision.
  • Lightweight and comfortable for extended wear.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Uncoated
Lens Tint Clear
Product Type Lens Insert
Frame Color Transparent
Material Plastic
Disposable Reusable
Lens Material Plastic
UV Protection None
Frame Type Frameless