Learn More
ERLAB Segmented Worksurface
Product Code. 30484794 Shop All Erlab Products
Change view
Click to view available options
For Use With (Equipment):
SOLIS 1200
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
30484794 SOLIS 1200 Each
1 options
Product Code. 30484794 Supplier ERLAB Supplier No. NORT000480
  • ONLINE EXCLUSIVE PRICE
    106.00 €
    98.00 € / Pack of 1
    Save 8.00 €
    7% Off
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Enhances flexibility and ease of maintenance in safety cabinets, designed for modular use
  • The Segmented Worksurface enhances flexibility
  • Ease of maintenance in safety cabinets
  • Designed for modular use
  • Allows partial removal for cleaning or setup changes
Specifications
Product Type Workstation
Housing Material Segmented Stainless Steel
For Use With (Equipment) SOLIS 1200