Learn More
Erlab Segmented Worktop
Product Code. 31794159 Shop All Erlab Products
Change view
Click to view available options
For Use With (Equipment):
Solis 1200
Solis 1500
Solis 1800
Solis 900
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31794159 Solis 900 Each
31794019 Solis 1200 Each
31794001 Solis 1500 Each
31794098 Solis 1800 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31794159 Supplier Erlab Supplier No. NORT000412
  • ONLINE EXCLUSIVE PRICE
    147.00 €
    137.00 € / Pack of 1
    Save 10.00 €
    6% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The worktop makes cleaning easier, due to a design that minimises areas where dirt can accumulate and improves access to surfaces.
  • Laboratory Worktops and Weighing Surfaces: Erlab worktops are designed to provide a durable, chemically resistant and easy-to-clean working surface for ductless enclosures, filtering storage cabinets and laboratory benches. Each surface is dimensioned to fit the corresponding Erlab equipment exactly.
  • Materials Selected for Laboratory Use
    • High-resistance materials suitable for daily contact with laboratory chemicals
    • Smooth, non-porous surface that is quick and easy to decontaminate
    • Resistant to scratching, impact and routine cleaning agents
    • Stainless Steel versions available for applications requiring maximum chemical and biological resistance
    • Segmented and weighing versions available for specific handling requirements
  • Designed for Safe Handling: The worktop geometry contributes to the containment performance of the enclosure by maintaining correct airflow at the working surface. Segmented versions allow easy removal for cleaning or for passing equipment through, while weighing surfaces provide a stable, vibration-damped supPort for precision balances.
  • Key Benefits:
    • Chemically resistant, easy-to-clean working surface.
    • Exact fit with the corresponding Erlab equipment.
    • Long service life under intensive laboratory use.
    • Options available for weighing and segmented configurations.
    • Contributes to correct airflow and containment performance.
Specifications
Product Type Flat Worksurface
For Use With (Equipment) Solis 900