missing translation for 'onlineSavingsMsg'
Learn More

Selenium, 99+%, powder

Product Code. 10764014 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10764014 500 g Glass bottle 500g
10289700 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10764014 Supplier Thermo Scientific Acros Supplier No. 419275000
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7782-49-2 | Se | 78.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-49-2
Molecular Formula Se
Molecular Weight (g/mol) 78.97
MDL Number MFCD00134090 MFCD00011224
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
Synonym selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray
PubChem CID 6326970
ChEBI CHEBI:27568
IUPAC Name selenium
SMILES [Se]

Specifications

Boiling Point 685°C
Melting Point 217°C
Color Cream-White to White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99% min
Linear Formula Se
Fieser 01,990; 04,422; 05,575; 06,507; 08,439; 10,345; 11,465; 14,281
Merck Index 15, 8568
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in concentrated acid,slightly soluble in diehtyl ether,almost insoluble in carbon disulfide,insoluble in ethanol
Formula Weight 78.96
Percent Purity ≥99%
Packaging Glass bottle
Residue after Ignition 0.5% max
Specific Gravity 4.81
Density 4.8100g/mL
Chemical Name or Material Selenium, <325 mesh
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Selenium
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H413-May cause long lasting harmful effects to aquatic life.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.