missing translation for 'onlineSavingsMsg'
Learn More

Selenium, Extra Pure, SLR, Fisher Chemical

Product Code. 10569010 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10569010 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10569010 Supplier Fisher Chemical Supplier No. S/0441/46
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 690°C
Melting Point 220°C
CAS 7782-49-2
Color Black to Gray
Physical Form Solid
Quantity 25 g
Molecular Formula Se
MDL Number MFCD00134090 MFCD00011224
Synonym selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
SMILES [Se]
IUPAC Name selenium
Molecular Weight (g/mol) 78.97
PubChem CID 6326970
ChEBI CHEBI:27568
Formula Weight 78.96g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Selenium
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H413-May cause long lasting harmful effects to aquatic life.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.