missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selexipag-d7 (iso-propyl-d7), CDN
Click to view available options
Quantity:
0.005 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Selexipag-D7 (N-isopropyl-D7) |
| CAS | 1265295-21-3 |
| Molecular Formula | C26 D7 H25 N4 O4 S |
| Quantity | 0.005 g |
| Synonym | Acetamide, 2-[4-[(5,6-diphenyl-2-pyrazinyl)[1-(methyl-d3)ethyl-1,2,2,2-d4]amino]butoxy]-N-(methylsulfonyl)- (ACI), 2-[4-[(5,6-Diphenyl-2-pyrazinyl)[1-(methyl-d3)ethyl-1,2,2,2-d4]amino]butoxy]-N-(methylsulfonyl)acetamide (ACI), Selexipag-d7, Selexipag-D7 (N-isopropyl-D7) |
| InChI Formula | InChI=1 S/C26H32N4O4S/c1-20(2)30(16-10-11-17-34-19-24(31)29-35(3,32)33)23-18-27-25(21-12-6-4-7-13-21)26(28-23)22-14-8-5-9-15-22/h4-9,12-15,18,20 H,10-11,16-17,19H2,1-3H3,(H,29,31)/i1D3,2D3,20 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(N(CCCCOCC(=O)NS(=O)(=O)C)c1cnc(c2ccccc2)c(n1)c3ccccc3)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-[4-[(5,6-diphenylpyrazin-2-yl)-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)amino]butoxy]-N-methylsulfonylacetamide |
| Molecular Weight (g/mol) | 503.6648 |
| Formula Weight | 503.2584 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction