missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selexipag
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
6243Selective prostacyclin IP1 receptor agonist (Ki values are 260 and 2100 nM and for human and rat IP receptors, and >10 μM for EP1-4, DP, FP, and TP receptors). Prodrug for the active form of MRE 269. Ameliorates vascular endothelial dysfunction and increases femoral skin blood flow in rats. Orally available.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 496.62 |
| Percent Purity | >98% |
| Chemical Name or Material | NS 304 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction