missing translation for 'onlineSavingsMsg'
Learn More

Semicarbazide hydrochloride, 99+%

Product Code. 11652209 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11652209 2.5 kg Plastic bottle 2.5kg
10265230 100 g Plastic bottle 100g
10061410 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11652209 Supplier Thermo Scientific Acros Supplier No. 132640025
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 563-41-7 | CH6ClN3O | 111.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-41-7
Molecular Formula CH6ClN3O
Molecular Weight (g/mol) 111.53
MDL Number MFCD00013009
InChI Key XHQYBDSXTDXSHY-UHFFFAOYSA-N
Synonym semicarbazide hydrochloride, aminourea hydrochloride, semicarbazide hcl, hydrazinecarboxamide hydrochloride, hydrazinecarboxamide, monohydrochloride, semicarbazide chloride, amidourea hydrochloride, carbamylhydrazine hydrochloride, semicarbazide monohydrochloride, semicarbazide, monohydrochloride
PubChem CID 11236
ChEBI CHEBI:82532
IUPAC Name aminourea;hydrochloride
SMILES [H+].[Cl-].NNC(N)=O

Specifications

Melting Point 170.0°C to 178.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula H2NCONHNH2·HCl
Quantity 2.5 kg
Beilstein 03,98
Fieser 01,1000; 15,284
Merck Index 15,8583
Solubility Information (10% in water) Clear colorless
Formula Weight 111.53
Percent Purity 99+%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Semicarbazide hydrochloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Semicarbazide hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Semicarbazide hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.