missing translation for 'onlineSavingsMsg'
Learn More

Semicarbazide Hydrochloride, TRC

Product Code. 30138132
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138132 5 g 5g
18070430 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138132 Supplier Toronto Research Chemicals Supplier No. TRCS2582005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-41-7
Molecular Formula C H5 N3 O . Cl H
Molecular Weight (g/mol) 111.53
Synonym Hydrazinecarboxamide, hydrochloride (1:1), Hydrazinecarboxamide, monohydrochloride (9CI), Semicarbazide, monohydrochloride (8CI), Aminourea hydrochloride, Hydrazinecarboxamide, hydrochloride, Semicarbazide chloride, Semicarbazide hydrochloride
IUPAC Name aminourea;hydrochloride
SMILES Cl.NNC(=O)N

Specifications

Quantity 5 g
InChI Formula InChI=1S/CH5N3O.ClH/c2-1(5)4-3;/h3H2,(H3,2,4,5);1H
Formula Weight 111.0199
Percent Purity >95%
Chemical Name or Material Semicarbazide hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.