Learn More
Semperit™ Semperguard™ Nitrile Comfort Powder-free Gloves
Product Code. 15594456
Change view
Click to view available options
Size:
9/10
Unit Size:
Pack of 90
Product Code. Size Unit Size
15594456 9/10 Pack of 90
1 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15594456 Supplier Semperit™ Supplier No. 3000001646
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Disposable protective glove made of Nitrile. Semperit™ Semperguard™ Nitrile Comfort Powder-free Gloves are latex free with high wearing comfort.
  • CE Category III protective glove for complex risks. CE Class I examination glove.
  • Special nitrile formulation with optimized resistance to oils and fats, good tear-resistance
  • Excellent wearing comfort
  • Tested and certified according to 1935/2004 - suitable for food contact
  • The problem-solver for single use to protect hands against chemical splashing and in the processing of fatty foods.
  • Free from phthalates / softeners and allergenic latex proteins
  • Signal color blue – suitable for areas subject to HACCP standards
Specifications
Color Blue
Material Nitrile
Powdered Powder Free
Size 9/10
Thickness (English) 7.8 mil
Thickness (Metric) 0.2 mm
Glove Cuff Style Rolled
Model 3000004109
Length (Metric) 240 mm