Learn More
SemperIt™ Semperguard™ Nitrile Xtra lite Protective Gloves
Product Code. 11817202
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 180
Pack of 200
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11817202 X-Large Pack of 180
11807202 Large Pack of 200
11897192 Medium Pack of 200
11887192 Small Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11817202 Supplier SemperIt™ Supplier No. 3000001620
  • ONLINE EXCLUSIVE PRICE
    32.00 €
    12.02 € / Pack of 180
    Save 19.98 €
    62% Off
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from nitrile. Semperguard™ Nitrile Xtra lite Protective Gloves are disposable.
  • Nitrile
  • Powder free
  • Lavender blue
  • Disposable
  • Rolled Cuff Style
  • Ambidextrous
  • Latex Free
  • Tensile Strength of 14/14MPa
  • CE Class I, EN 420, EN 374, EN 455, PPE Category III certified 
Specifications
Certifications/Compliance CE Class I, EN 420, EN 374, EN 455, PPE Category III
Color Purple
Disposable Single-use
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Large
Thickness (English) 5.5 mil
Thickness (Metric) 0.14 mm
Glove Cuff Style Rolled
Model 816780239
Tensile Strength 14 MPa
Length (Metric) 240 mm