missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Comfort Latex Examination Gloves

Product Code. 15303884
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 100
Pack of 90
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15303884 Small Pack of 100
15383874 Large Pack of 100
15313884 X-Large Pack of 90
15393874 Medium Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15303884 Supplier Semperit™ Supplier No. 3000001280

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Semperguard™ Comfort Latex Examination Gloves are powder-free and provide excellent elasticity and are very comfortable to wear.

  • Made of natural latex
  • Skin-friendly
  • Powder-free
  • Excellent elasticity and good tactility/ wearing comfort
  • Good grip and easy to don
  • Free from phthalates and softeners
  • CE category Medical device class ISingle-use protective glove for complex risks category III
  • Tested and certified according to 1935/2004
  • Suitable for food contact

Specifications

Certifications/Compliance EN 420, EN 374 and EN 455
Classification CE Class I
Color White
Disposable Single-use
Laboratory/Cleanroom Use Yes
Latex Free No
Material Latex
Powdered Powder Free
Product Type Examination Gloves
Size Small
Sterility Non-sterile
Thickness (Metric) 0.20 mm
Finger Style Textured
For Use With (Application) Work Safety, Care, Laboratory/Pharma, Cleaning/Hygiene, Food, Retail/Household
Glove Cuff Style Rolled
Inspected for Medical Use Yes
Packaging Type Dispenser Box
Quantity 100 Pk.
Resistant To Chemical
Length (Metric) 240 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.