missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Style Nitrile Gloves

Produktkod. 15583536
missing translation for 'orderingAttributeHoverText'
Size:
Small
Medium
Large
Förpackningsstorlek
Pack of 100
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 15583536

missing translation for 'mfr': Semperit™ 3000003331

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

The Black Nitrile Glove. Semperit™ Semperguard™ Style Nitrile Gloves are non-sterile, unpowdered, latex-free black nitrile gloves with excellent stretchability, a rolled cuff for easy donning and textured fingers for improved grip.

  • Black nitrile glove with excellent stretchability
  • Particularly suitable for front-end applications in the catering, beauty and body art sector
  • Thanks to dual labeling, also meets the medical requirements of the tattoo and piercing sector
  • Free from phthalates / plasticizers and allergenic latex proteins
  • Rolled cuff enables easy donning
  • Convenient dispenser box holds 100 ambidextrous gloves
  • CE category III protective glove for complex risks, CE class I examination glove
  • Tested and certified according to 1935/2004 - suitable for food contact

Specifikationer

Certifications/Compliance Conforms to standards EN 420, EN 374, EN455
Classification CE Category III Protective Glove for Complex Risks, CE Class I Examination Glove 
Color Black
Disposable Single-use
Exterior Finish Fingertip Textured
Grade Certified according to VO 1935/2004 - suitable for contact with food
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Medium
Sterility Non-sterile
Thickness (Metric) 0.14 mm
For Use With (Application) Especially suitable for applications which are visible to the public
Glove Cuff Style Beaded/Rolled
Model Style Nitrile
Packaging Type Dispenser box
Quantity 100/bx
Length (Metric) 240 mm
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.