missing translation for 'onlineSavingsMsg'
Learn More

SemperIt™ Semperguard™ Powder-Free Disposable Vinyl Gloves

Catalog Number 12201338
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured from a high quality vinyl formula, Semperguard™ Powder-Free Disposable Vinyl Gloves are tear-resistant, seamless gloves that are flat with a rolled edge.

Disposable transparent protective gloves are flat with a rolled edge.
  • Particularly skin-friendly for latex allergy sufferers - free of latex proteins and accelerators
  • Free of DOP (DEHP / diethylhexyl phthalate)
  • High quality vinyl (PVC) provides excellent wearing comfort
  • Excellent quality at an economical cost
  • CE category I Protective glove for minimal risks
  • Tested and certified according to VO 1935/2004 - suitable for contact with non-fatty foods
Certifications/Compliance CE Category I, EN 420, EN 374, EN 455
Classification CE category I Protective glove for minimal risks
Color Transparent
Disposable Single-use
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Vinyl
Powdered Powder Free
Size X-Large
Sterility Non-sterile
Thickness (Metric) 0.14 mm
Glove Cuff Style Rolled
Packaging Type Dispenser box
Palm Type Flat
Length (Metric) 240 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'