missing translation for 'onlineSavingsMsg'
Learn More

SemperIt™ Semperguard™ Powder-Free Disposable Vinyl Gloves

Product Code. 12312189
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 100
Pack of 90
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12312189 Large Pack of 100
12302189 Medium Pack of 100
12392179 Small Pack of 100
12201338 X-Large Pack of 90
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12312189 Supplier SemperIt™ Supplier No. 3000001446
  • ONLINE EXCLUSIVE PRICE
    15.59 €
    4.55 € / Pack of 100
    Save 11.04 €
    70% Off
View all promotions
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from a high quality vinyl formula, Semperguard™ Powder-Free Disposable Vinyl Gloves are tear-resistant, seamless gloves that are flat with a rolled edge.

Disposable transparent protective gloves are flat with a rolled edge.
  • Particularly skin-friendly for latex allergy sufferers - free of latex proteins and accelerators
  • Free of DOP (DEHP / diethylhexyl phthalate)
  • High quality vinyl (PVC) provides excellent wearing comfort
  • Excellent quality at an economical cost
  • CE category I Protective glove for minimal risks
  • Tested and certified according to VO 1935/2004 - suitable for contact with non-fatty foods

Specifications

Certifications/Compliance CE Category I, EN 420, EN 374, EN 455
Classification CE category I Protective glove for minimal risks
Color Transparent
Disposable Single-use
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Vinyl
Powdered Powder Free
Size Large
Sterility Non-sterile
Thickness (Metric) 0.14 mm
Glove Cuff Style Rolled
Packaging Type Dispenser box
Palm Type Flat
Length (Metric) 240 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.