missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SEN 177
GREENER_CHOICE

Product Code. p-7223021
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18758736

Brand: Tocris Bioscience™ 6979/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent glutaminyl cyclase inhibitor; disrupts binding of SIRP-α to CD47

SEN 177 is a potent glutaminyl cyclase (QPCT) inhibitor (IC50 = 53 nM). SEN 177 reduces aggregation of mutant huntingtin (mHTT) through inhibition of QPCT and reduces Q80 aggregates in vitro. Inhibition of QPCT also reduces aggregate formation in in vivo models of Huntington's disease. SEN 177 potently inhibits QPCT-like protein (IC50 = 13 nM) and disrupts binding of SIRP-α to CD47. The compound increases antibody-dependent cellular phagocytosis and cytotoxicity in cancer cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6 '-Fluoro-2-[4-(4-methyl-4 H-1,2,4-triazol-3-yl)-1-piperidinyl]-3,3 '-bipyridine
CAS 2117405-13-5
Quantity 10 mg
Target Other Transferase Inhibitors
Molecular Formula C18H19FN6
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.