missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Sensilite™ 48-100 Series White Polyurethane Light Weight Gloves

Product Code. 12660912
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12660912 10 12 pairs
12670912 11 12 pairs
12680912 6 12 pairs
12690912 7 12 pairs
12600922 8 12 pairs
12610922 9 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 12660912 Supplier Ansell™ Supplier No. 48100100
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed for light applications that require good levels of comfort and abrasion resistance.

  • Industrial gloves
  • Abrasion resistance for extended wear life
  • Thin liner and coating offering great dexterity and flexibility
  • Sensilite 48-100 allows extreme tactility where precision is required
  • White colour to ensure process quality and avoid cross contamination
  • Comfort and protection for lightweight applications
  • Offers precise handling of small objects in dry or light oily environments
  • Provides enhanced safety, dexterity and durability

Applications:

  • Assembly and disassembly of equipment
  • Chassis assembly, body trimming
  • Equipment repair and maintenance
  • Loading and unloading rough parts or boxes
  • Picking parts and accessories
  • Production line support and maintenance
TRUSTED_SUSTAINABILITY

Specifications

Back Material Polyurethane
Certifications/Compliance Standard CATEGORY II EN 388 : 4131
Cuff Material Nylon
Coating Material Polyurethane
Color White
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Polyurethane
Product Type Lightweight Gloves
Size 10
Glove Cuff Style Knit
Liner Material Nylon
Lining Lined
Palm Type Coated-palm
Quantity 12 Pairs
Resistant To Abrasion resistance
Series 48-100
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.