missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sentino™ Magnetic Filters Funnel

Product Code. p-7089780
Change view
Click to view available options
Capacity (Metric):
150 mL
300 mL
500 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17174291 300 mL Each
17164291 150 mL Each
17184291 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17174291 Supplier Cytiva Supplier No. 4273
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Magnetic funnels designed for use with our Sentino pump.

  • Unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes
  • Not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric)Top 47 mm
Capacity (Metric) 300 mL
Material PPSF
Autoclavable Autoclavable
Description 9.6 cm2
Product Type Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.