missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Septa, 3-Layer, 9.5 mm DIA (12/PK)

Product Code. 11592974
Change view
Click to view available options
Diameter (Metric) Inner:
9.5 mm
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Unit Size
11592974 9.5 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11592974 Supplier Hamilton™ Supplier No. 76004
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Three Layer Septa is made of preconditioned silicone rubber and is 3.81 mm (0.150 in) thick.

212°C / 414°F

Specifications

Product Type 3-Layer
Diameter (Metric) Inner 9.5 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.