missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Septa Cathode Buffer Container, for 3500 and SeqStudio™ Flex

Product Code. 12618343 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
10 Containers
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12618343 10 Containers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12618343 Supplier Applied Biosystems™ Supplier No. 4410715
Black Friday Starts Here
Use promo code 27984
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These septa are for both compartments of the Cathode Buffer Container, for use with the 3500 series Genetic Analyzers.

These septa are for both compartments of the Cathode Buffer Container and are for use with 3500 and SeqStudio Flex series genetic analyzers.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 Containers
For Use With (Equipment) 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
Product Type Septa Cathode Buffer Container
Shipping Condition Room Temperature
Content And Storage This product contains 10 septa.

For Research Use Only. Not for use in diagnostic procedures.

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.