missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Septa for 8-Well Strips, for 3500/SeqStudio™ Flex

Product Code. 15809856
Change view
Click to view available options
Quantity:
24 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15809856 24 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15809856 Supplier Applied Biosystems™ Supplier No. 4410701
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These Septa for 8-strips were designed and validated for use with Applied Biosystems 3500 and SeqStudio Flex Series Genetic Analyzers.

These septa for 8-tube strips were designed and validated for use with Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 24 each
Format 8-well Septa Strip
For Use With (Equipment) 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
Product Type Septa Strip
Shipping Condition Room Temperature
Content And Storage This product contains 24 septa.
When I started my run on the Applied Biosystems 3500/3500xL Genetic Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?

Some of the other causes of an Unstable Electrophoresis current detected error message are:

1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues

Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.

Do I need to replace the buffer every 14 days if the Applied Biosystems 3500/3500xL Genetic Analyzer is not in use?

If the instrument is not in use, it is not necessary to replace the buffer every 14 days. However, the buffer level needs to remain at the fill line to prevent the capillary or array from drying out. Top off the buffer volume with water if necessary. Replace the anode and cathode buffer containers prior to starting a new run.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.