Learn More
Applied Biosystems™ SeqScape™ Software v3.0, Initial License
Product Code. 15908206
Change view
Click to view available options
Quantity:
1 license
Unit Size:
Pack of 3
Product Code. Quantity Unit Size
15908206 1 license Pack of 3
1 options
Product Code. 15908206 Supplier Applied Biosystems™ Supplier No. 4474978
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This is an Initial License for the SeqScape™ Software.
The SeqScape™ Software is a resequencing package designed for mutation detection and analysis, SNP discovery and validation, pathogen sub-typing, allele identification, and sequence confirmation. It provides library functions for comparison to a known group of sequences, as well as 21 CFR Part 11 functionalities.
TRUSTED_SUSTAINABILITY
Specifications
Product Line SeqScape
Software Type Sequencing Data Analysis
Content And Storage Software/CD
Data Entry CE Sequencing (.ab1 files)
License Initial
Operating System Windows 7 Pro
Product Type SeqScape Software V3.0 Initial License
Quantity 1 license
Software Category Supports files from all AB Genetic Analyzers