Learn More
Applied Biosystems™ SeqScape™ Software v4.0, Initial License
Description
- This is an Initial License for SeqScape Software
- SeqScape Software is a resequencing package designed for mutation detection and analysis, SNP discovery and validation, pathogen sub-typing, allele identification, and sequence confirmation
- It provides library functions for comparison to a known group of sequences, as well as 21 CFR Part 11 functionalities
Specifications
Specifications
| Product Line | SeqScape |
| Software Type | Sequencing Data Analysis |
| Content And Storage | Software/CD |
| Data Entry | CE Sequencing (.ab1 files) |
| License | Initial |
| Operating System | Windows 10 Pro/IoT, Windows 7 Pro |
| Product Type | SeqScape Software V4.0 Initial License |
| Quantity | 1 license |
| Software Category | Supports files from all AB Genetic Analyzers |