missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Sequence Detection Primer

Product Code. 15592413 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
10,000 pmol
130,000 pmol
80,000 pmol
Unit Size:
130000pmol
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15592413 130,000 pmol 130000pmol
12015030 10,000 pmol Each
10565677 80,000 pmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15592413 Supplier Applied Biosystems™ Supplier No. 4304972
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sequence Detection Primer

These unlabeled primers can be used with TaqMan™ probes in any real-time PCR application. Choose between dry and in-solution formatted delivery. All primers are desalted.

Sequence detection primers are unlabeled primers that can be used with TaqMan probes or SYBR Green dye for your real-time PCR research applications.

Features include:

  • Choice of delivered scales
  • For use in any real-time PCR or PCR application
  • All sequence detection primers are desalted

Real-time PCR sequence detection primers are delivered in either dry or liquid format.

Guaranteed yield

The unit size you select is based on the minimum amount we will deliver, not on synthesis starting amount.

Approximate ship time

4–6 days in North America, 5-10 days in Europe

We also offer:

  • Custom TaqMan probes with a variety of options for dyes and quenchers.
  • TaqMan MGB probes and primers available as predesigned and pre-formulated TaqMan Assays.
  • Commercial manufacturing products and services, including OEM and GMP oligo manufacturing.

Need a different dye or concentration or a larger lot size? The TaqMan Custom Assay and Oligo Service offers custom products that you can tailor to meet your experimental needs.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex, QuantStudio™ Absolute Q Digital PCR System
Product Type Sequencing Primer
Content And Storage 1 tube containing an unlabeled primer in solution or dried down.

Store at -15°C to -25°C
Purification Method Desalted
Shipping Condition Room Temperature
Primer Unlabeled
Quantity 130,000 pmol
Product Line Applied Biosystems
For Use With (Application) Real Time PCR Applications
Form Liquid or Dried Down

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.