missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Sequence Detection System v2.4 Enterprise Edition

Product Code. 15830363 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 software
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15830363 1 software Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15830363 Supplier Applied Biosystems™ Supplier No. 4350490
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sequence Detection System (SDS) Software v2.

Features of SDS Software:

  • Assisted Tm Calling
  • Ability to create and save analysis templates on a per detector basis for use with other plates
  • Auto-analysis of plates containing analysis settings
  • Improvements in amplification plot displays, including the ability to view plots by sample and target, as well as a toggle for linear/log display
  • Ability to access the plate utility application from the main menu
  • Ability to export the results or data of up to 50 plates at once, as well as a choice of results columns to export, using the batch export utility
  • Ability to set up standard curve dilutions for AQ experiments with a wizard
  • Ability to switch run documents from AQ to RQ format, and vice-versa
Standard and Enterprise Editions
The standard version of SDS Software for the 7900HT Fast Real-Time PCR System uses two automation software tools—Plate Utility and Automation Controller—to fully automate high-throughput applications for gene expression and SNP genotyping, requiring minimal user intervention.

The Enterprise Edition of SDS Software is a high-throughput application for storage, organization, and analysis of large amounts of gene expression and genotyping data on the 7900HT Fast Real-Time PCR System
  • Upgrade to Oracle Database 11g
  • Process-based quality flags for increased quality control, problem notification, filtering, and troubleshooting
  • Improved review and data group tools
  • Wizard-based plate document creation
  • Support for plus/minus assays
  • Re-engineered SDS Automation Controller
  • Ability to save plates created using the template batch utility directly to the database
The Enterprise database allows users to process up to 5,000 x 384-well plates (384 markers total), which equates to 1.92 million genotypes. Using Relative Quantitation (RQ) Manager, customers can also process 200 x 384-well plates (96 detectors with quadruplicate data points per multiplexed endogenous control) which equates to 153,600 data points.

Supporting 21 CFR Part 11 Requirements
SDS Software Enterprise Edition is the only edition of SDS Software that assists in supporting 21 CFR part 11 requirements, providing security, auditing functions, electronic signatures, and electronic record-keeping. These features eliminate traditional paper records and reduce vulnerability to signature fraud and report misfiling, which increases the speed of information exchange and data searching. The results are reduced storage costs, better data integration, and superior product quality.

For Research Use Only. Not intended for any animal, human therapeutic or diagnostic use.
TRUSTED_SUSTAINABILITY

Specifications

Software Type Real-Time PCR Data Analysis
Content And Storage 1 Software Kit
Format 384-Well Plate
For Use With (Equipment) Applied Biosystems 7900HT Fast Real Time PCR System
Product Type Sequence Detection System (SDS) Software
Quantity 1 software
Software Category Software
What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?

Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.