missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sequential Access Storage Option for the Orbitor™ RS2 Microplate Mover for QuantStudio™ Real-time PCR Systems

Product Code. 17162504 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity unitSize
17162504 1 Ea. Each
1 options
This item is not returnable. View return policy

Product Code. 17162504

Brand: Thermo Scientific™ A43781

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for use with the Orbitor RS2 Microplate Mover and a QuantStudio real-time PCR system, enabling real-time PCR in automated environments for increased sample throughput

  • This Sequential Access Storage Option is designed for use with the Orbitor RS2 Microplate Mover and a QuantStudio real-time PCR system, enabling real-time PCR in automated environments for increased sample throughput.
  • This storage option holds up to 20 plates/3 stacks (96-well) and up to 40 plates/3 stacks (384-well). These stacks provide the option for the Orbitor RS2 mover to run until empty, which allows for additional plates to be added while the Orbitor RS2 mover is running. This option is ideal for 0.1-mL and 0.2-mL 96-well plates and 384-well plates.
  • Couple this Sequential Access Storage Option with the Orbitor RS2 Microplate Mover for QuantStudio Systems to complete your automation bundle.
  • For integration of two QuantStudio systems with the Orbitor RS2 mover, the Dual QuantStudio System Integration Kit is required in addition to the automation bundle.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) QuantStudio 7 Pro, QuantStudio 7 Flex, or QuantStudio 12 K Flex Real-Time PCR Systems and The Orbitor RS2 Microplate Mover
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Room Temperature
Description Storage Option
Quantity 1 Ea.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.