missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Sequential DNase Buffer, 100 rxns

Product Code. 31781085 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Vial
Unit Size:
96 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31781085 1 Vial 96 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31781085 Supplier Applied Biosystems™ Supplier No. A40004573

Please to purchase this item. Need a web account? Register with us today!

Sequential DNase Buffer is a component of the Sequential Protein/DNA/RNA Extraction Kit (Cat. No. 50000D) used for RNA preparation.

Applied Biosystems Sequential DNase Buffer is a component of the Sequential Protein/DNA/RNA Extraction Kit used for RNA preparation.

Features of the Sequential Protein/DNA/RNA Extraction Kit include:

  • High sensitivity—supports scalable extractions from precious or low sample volumes down to 1,000 cells or 0.005 mg tissue (tissue dependent)
  • Direct compatibility—extracted analytes down to 100,000 cells or 1-2 mg tissue are validated and suitable for mass spectrometry and NGS applications
  • Automated high-throughput—compatible with KingFisher instruments for scalable processing
  • Enhanced reproducibility—standardized protocol reduces variability

The kit employs a bead-based sequential extraction methodology utilizing protein isolation prior to nucleic acid extraction, resulting in enhanced protein yields that are directly compatible with mass spectrometry analysis. This approach enables researchers to obtain analytes suitable for diverse downstream applications including SDS-PAGE, western blotting, mass spectrometry, sequencing, genotyping, and qPCR from sample inputs as small as 1-2 mg of tissue or 100,000 cells while maintaining quality standards required for both next-generation sequencing (NGS) and MS platforms. The Dynabeads and MagMAX magnetic bead-based purification technology helps enable high reproducibility and scalable, high-throughput processing when integrated with automation systems such as the KingFisher Sample Purification System.

This kit combines the advantages of multi-analyte extraction with the precision and efficiency required for advanced proteomics and genomics workflows, making it particularly valuable for multi-omics studies where sample conservation and analytical comprehensiveness are paramount.

Verified performance

The kit helps deliver yield and quality metrics comparable to standard single-analyte extraction kits, facilitating efficient multi-analyte recovery from a single sample. While most commercial sequential protocols are optimized for large sample volumes (e.g. 1 million cells or 10–20 mg tissue), this kit offers exceptional versatility across a wider range. Validated for 100,000 cells and 1–2 mg tissue, it has also been successfully used with as few as 1,000 cells and 5 μg tissue while maintaining linear yields and preserving nucleic acid integrity suitable for qPCR analysis. This broad dynamic range provides greater flexibility and enables efficient processing of precious or limited samples that are difficult to analyze with conventional methods.

Streamlined three-step sequential extraction

The extraction procedure follows a systematic three-step process designed to support a high recovery of each biomolecule class. Initially, Dynabeads Protein Beads are introduced to the lysed sample and incubated to facilitate protein binding, after which they are transferred to a separate tube for washing and protein processing compatible with downstream western blot or mass-spectrometry applications. Subsequently, MagMAX DNA/RNA Binding beads are added to the original tube, now depleted of proteins, to enable DNA binding and extraction using proprietary buffer formulations before transferring to a new tube for further preparation. Finally, MagMAX DNA/RNA Binding beads are again introduced to the original tube, now depleted of both proteins and DNA, to facilitate RNA binding and extraction using specialized proprietary buffers and solutions, with subsequent transfer to a dedicated tube for final preparation. The resulting extracted proteins are suitable for WB or MS analysis, while the recovered DNA and RNA are compatible with quantitative PCR (qPCR) and next-generation sequencing (NGS) applications.

TRUSTED_SUSTAINABILITY

Specifications

Bead Type DNase Buffer
Capacity 1 mL
Content And Storage •The vial contains DNase Buffer used in the RNA purification step.
•Store at room temperature.
Format Reagent
Isolation Technology Magnetic Bead
Sample Type Cell Lines, PBMC, Fresh/Frozen Tissue
Formulation Reagent Solution
For Use With (Equipment) KingFisher™ Duo Prime, Flex, or Apex Purification Systems
For Use With (Application) Mass spectrometry, western blot, qPCR, sequencing
No. of Reactions 100 Reactions
Product Line Dynabeads, MPC
Product Type Sequential Protein/DNA/RNA Extraction
Quantity 1 Vial
Shipping Condition Ambient Temperature
Throughput Flexible
Regulatory Status For Research Use Only
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.