missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag™ Carboxylate modified Magnetic Beads, hydrophobic

Product Code. 13273589 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
15 mL
100 mL
1 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13273589 15 mL Each
11879782 100 mL Each
11889782 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13273589 Supplier Cytiva Supplier No. 44152105050250
  • ONLINE EXCLUSIVE PRICE
    434.00 €
    396.80 € / Pack of 1
    Save 37.20 €
    8% Off
View all promotions
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Excellent for coupling oligos or proteins directly to beads. Their cauliflower-like structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these single shell magnetite beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.

  • High binding capacity
  • 1.0μm diameter
  • High carboxyl acid content for binding for binding DNA and proteins
  • Hydrophobic bead based on crosslinking requirements for the encapsulation layer
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5% solids (50mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) DNA Sample Prep, Protein Purification
Quantity 15 mL
Diameter (Metric) 1 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.