missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag SpeedBeads Amine-Blocked Particles

Product Code. 11834962 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
Each
Product Code. Quantity unitSize
11834962 5 mL Each
1 options
This item is not returnable. View return policy

Product Code. 11834962

Brand: Cytiva 19152104010150

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reduce undesired protein adsorption. Cytiva Sera-Mag SpeedBeads Amine-Blocked Particles respond twice as fast and feature a non-surfactant, non-protein blocking reagent. They are the ideal choice for molecular biology, nucleic acid isolation, research and clinical diagnostic immunoassays.

Sera-Mag SpeedBeads Amine-Blocked Particles combine a fast magnetic response time and high binding capacity with a large surface area, high sensitivity, stability, physical integrity, and fast reaction kinetics.
  • Non-surfactant, non-protein blocking reagent provides very low non-specific binding of proteins from the sample matrix
  • Fast reaction kinetics increases throughput and precision, and also enables faster movement through viscous solutions
  • Unique cauliflower-like surface provides increased area for binding reactions compared to smooth surface particles
  • Uniform, nominal 1μm diameter provides excellent lot-to-lot reproducibility
Sera-Mag SpeedBeads Respond Twice As Fast
The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer. Two layers of magnetite are coated onto this core, resulting in faster magnetic response times. The surface is then blocked with a non-surfactant, non-protein based method, to help prevent non-specific binding of proteins
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10mg/ml
Content And Storage 4°C
Quantity 5 mL
Diameter (Metric) 1 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.