missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag SpeedBeads Amine-Blocked Particles

Product Code. 11844962 Shop All Cytiva Products
Change view
Click to view available options
Packaging:
1 mL
100 mL
Unit Size:
Each
Product Code. Packaging unitSize
11844962 100 mL Each
11854962 1 mL Each
2 options
This item is not returnable. View return policy

Product Code. 11844962

Brand: Cytiva 19152104010350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sera-Mag SpeedBeads Amine-Blocked Particles reduce undesired protein adsorption.

  • Combine a fast magnetic response time and high binding capacity with a large surface area, high sensitivity, stability, physical integrity, and fast reaction kinetics
  • Typical applications include molecular biology, nucleic acid isolation, research, and clinical diagnostic immunoassays
  • The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer
  • Two layers of magnetite are coated onto this core, resulting in faster magnetic response times
  • The surface is blocked with a non-surfactant, non-protein based method, to help prevent non-specific binding of proteins
  • TRUSTED_SUSTAINABILITY

    Specifications

    Binding Property Amine
    Concentration 10 mg/mL
    Material Magnetite
    Packaging 100 mL
    Diameter (Metric) 1 μm
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.