missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Their cauliflower-like structure gives Sera-Mag beads high binding capacity
- Fast and easy IgA purification and IgG purification
Specifications
Specifications
| Concentration | 1% solids (10mg/mL) |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Affinity Purification |
| Quantity | 1 mL |
| Diameter (Metric) | 1 μm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?