missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Ideal for immunoassay and molecular biology applications, including sample preparation and assay development for genomics and proteomics
- Available with various nominal biotin binding capacities for optimizing assay development
- Low dissociation constant to tightly bound ligands
- Low non-specific binding for greater precision and accuracy
- Even streptavidin coating for reliable results
- Uniform, nominal 1μm diameter provides high surface area and excellent lot-to-lot reproducibility
The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer. A layer of magnetite is then coated onto this core.
Specifications
Specifications
| Binding Property | 2500 to 3500 pmol/mg |
| Content And Storage | 4°C |
| Quantity | 5 mL |
| Diameter (Metric) | 1 μm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?