missing translation for 'onlineSavingsMsg'
Learn More

3M™ Series G3000 Slim Design Hard Hat with Leather Sweatband

Product Code. 15749987 Shop All 3M Products
Change view
Click to view available options
Color:
Hi-vis Yellow
White
Yellow
Unit Size:
Pack of 20
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15749987 White Pack of 20
15759987 Yellow Pack of 20
15789987 Hi-vis Yellow Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15749987 Supplier 3M™ Supplier No. 7000108322
  • ONLINE EXCLUSIVE PRICE
    924.00 €
    720.29 € / Pack of 20
    Save 203.71 €
    22% Off
Estimated Shipment: 26-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3M™ Series G3000 Slim Design Hard Hat with Leather Sweatband allows falling objects to slide off easily and not dislodge the helmet from its position.

  • A hard hat designed for use in harsh environments, with excellent ventilation and a good field of vision
  • 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV- radiation and its time to replace it
  • Softly rounded design, which helps to avoid helmet parts snagging on branches, etc
  • Head harness can be rotated 180° for better visibility when working in confined spaces
  • Short peak gives improved upwards visibility
  • Electrical insulation to 1000Vac
  • Easy to replace soft leather sweatband for the highest level of comfort and hygiene
TRUSTED_SUSTAINABILITY

Specifications

Color White
Includes Leather Sweatband
Material UV-stabilized ABS
Product Type Helmet
Size 54 to 62 cm
Weight (Metric) 330 g
Ventilated Not Ventilated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.