missing translation for 'onlineSavingsMsg'
Learn More

Kartell SpA Seropet

Product Code. 10244412
Change view
Click to view available options
Color:
Assorted
Red
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10244412 Assorted Each
10551583 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10244412 Supplier Kartell SpA Supplier No. 0136100
  • ONLINE EXCLUSIVE PRICE
    60.30 €
    57.15 € / Pack of 1
    Save 3.15 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pipette aspirator developed for aspirating liquids by glass or plastic pipettes ranging from the volume of 1 mL up to 100 mL.

The particularly light structure ensures comfortable use even when used for a long time and its particular configuration also makes it ideal for underhood uses. The buttons are very easy to access for choosing different modes depending on the volume pipette being used and viscosity of the liquid to aspirate. It is protected from cross contamination in the event of excessive aspiration by the filter on the aspiration device and the safety valve. It is supplied complete with battery charger, protective PTFE hydrophobic filter (0.2 μm) and an adhesive support for attachment to the laboratory bench.

Specifications

Autoclavable Autoclavable
Color Assorted
Disposable Reusable
For Use With Serological Pipettes
Grip Type Ergonomic
Material Polypropylene
Number of Channels 1
Sterility Non-sterile
Type Filler/Dispenser
Volume (Metric) 100 mL
Voltage 220 V
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.