missing translation for 'onlineSavingsMsg'
Learn More

Serotonin Creatine Sulfate Monohydrate(5-Hydroxy Tryptamine Creatinine Sulfate Monohydrate), TRC

Product Code. 30397066
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30397066 100 mg -
30389336 1 g 1g
30392548 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397066 Supplier Toronto Research Chemicals Supplier No. TRCH974990100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-47-2
Molecular Formula C10 H12 N2 O . C4 H7 N3 O . H2 O . H2 O4 S
Molecular Weight (g/mol) 405.43
Synonym 5-Hydroxytryptamine creatinine sulfate monohydrate
IUPAC Name 3-(2-aminoethyl)-1H-indol-5-ol;2-imino-1-methylimidazolidin-4-one;sulfuric acid;hydrate
SMILES O.CN1CC(=O)NC1=N.NCCc2c[nH]c3ccc(O)cc23.OS(=O)(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H12N2O.C4H7N3O.H2O4S.H2O/c11-4-3-7-6-12-10-2-1-8(13)5-9(7)10;1-7-2-3(8)6-4(7)5;1-5(2,3)4;/h1-2,5-6,12-13H,3-4,11H2;2H2,1H3,(H2,5,6,8);(H2,1,2,3,4);1H2
Formula Weight 405.1318
Chemical Name or Material Serotonin creatine sulfate monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.