missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Serotonin hydrochloride

Product Code. 30001419
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001419 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001419 Supplier R&D Systems™ Supplier No. 3547/50
  • ONLINE EXCLUSIVE PRICE
    150.40 €
    134.40 € / 50mg
    Save 16.00 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Endogenous 5-HT receptor agonist

Endogenous agonist at 5-HT receptors and endogenous substrate for 5-HT transporters. Neurotransmitter that has roles in regulation of mood, emesis, sexuality, sleep and appetite in vivo. Caged serotonin is also available (Cat.No. 3991).
TRUSTED_SUSTAINABILITY

Specifications

CAS 153-98-0
Quantity 50 mg
Product Line Tocris
Molecular Formula C10H13ClN2O
Formula Weight 212.68
Purity >99%
Molecular Weight (g/mol) 212.677
Chemical Name or Material Serotonin hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.