missing translation for 'onlineSavingsMsg'
Learn More

Serotonin hydrochloride, 97+%

Product Code. 10647012 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10647012 100 mg Glass bottle 100mg
10265921 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10647012 Supplier Thermo Scientific Acros Supplier No. 215021000
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 153-98-0 | C10H12N2O·HCl | 212.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153-98-0
Molecular Formula C10H12N2O·HCl
Molecular Weight (g/mol) 212.68
MDL Number MFCD00012686
InChI Key MDIGAZPGKJFIAH-UHFFFAOYSA-N
Synonym serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride
PubChem CID 160436
IUPAC Name 3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride
SMILES C1=CC2=C(C=C1O)C(=CN2)CCN.Cl

Specifications

Melting Point 168°C to 172°C
Color Yellow to Beige
Loss on Drying 2.0 % max. (115°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Quantity 100 mg
UN Number 2811
Beilstein 22, III, 5668
Merck Index 14, 8462
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
Formula Weight 212.68
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Serotonin hydrochloride, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Serotonin hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.