missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Serum Bottles

Product Code. p-7111358 Shop All Labconco Corporation Products
Click to view available options
Capacity (Metric):
10 mL
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 15113065

Brand: Labconco™ 7573210

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Perfect for long term storage of freeze dried samples. Labconco Serum Bottles are specifically designed for lyophilization applications.

  • Uniform thin wall construction ensures even freezing and drying
  • Ideal containers for use in the FreeZone Stoppering Tray Dryer
  • May be connected to valve ports on drying chambers and manifolds
  • Supplied in packages of 100

Specifications

Capacity (Metric) 10 mL
Color Clear
Material Glass
No. per Pack 100/PK
Sterility Non-sterile
Product Type Serum Bottle
Closure Size 20 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.