missing translation for 'onlineSavingsMsg'
Learn More

Sesame oil, MP Biomedicals™

Product Code. 11450961 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
Product Code. Quantity unitSize
11450961 250 mL 250mL
11460961 1 L 1 L
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11450961

Brand: MP Biomedicals 0215662183

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Chemical Identifiers

CAS 8008-74-0
Synonym Benne oil, Teel oil

Specifications

Density 0.915-0.925g/mL
Quantity 250 mL
Physical Form Oily Liquid
Chemical Name or Material Sesame oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.