missing translation for 'onlineSavingsMsg'
Learn More

Sesame oil, MP Biomedicals™

Product Code. 11460961 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460961 1 L 1 L
11450961 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460961 Supplier MP Biomedicals Supplier No. 0215662191
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 8008-74-0
Synonym Benne oil, Teel oil

Specifications

Density 0.915-0.925g/mL
Quantity 1 L
Physical Form Oily Liquid
Chemical Name or Material Sesame oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.