missing translation for 'onlineSavingsMsg'
Learn More

Sesame oil

Product Code. 10157453 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10157453 250 mL Glass bottle 250mL
10307990 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10157453 Supplier Thermo Scientific Acros Supplier No. 241002500
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 8008-74-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8008-74-0
MDL Number MFCD00132243

Specifications

Acidity 0.6mg KOH/g max.
Melting Point -5.0°C
Color Yellow
Density 0.9190g/mL
Flash Point 255°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4700 to 1.4760
Quantity 250 mL
Merck Index 14, 8469
Specific Gravity 0.919
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, ether, petroleum ethers
Physical Form Viscous Liquid
Chemical Name or Material Sesame oil
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.