missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Sesamol

Product Code. 16469156
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469156 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16469156 Supplier MedChemExpress Supplier No. HYN1417
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Sesamol is a constituent of sesame oil. Sesamol shows a free radical scavenging activity. Sesamol shows an IC50=5.95±0.56 μg/mL in the DPPH assay. Anti-oxidant activities. Anticancer activities.

Specifications

Content And Storage 4°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)
Molecular Weight (g/mol) 138.12 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 533-31-3
Product Type Natural Products
Purity 97.0%
Solubility DMSO : ≥ 100 mg/mL (724.01 mM) ∣Ethanol : ≥ 100 mg/mL (724.01 mM)
Target Apoptosis
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.