missing translation for 'onlineSavingsMsg'
Learn More

Sesamol, 98%

Product Code. 10048591 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10048591 5 g Glass bottle 5g
10193370 25 g Glass bottle 25g
10455175 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10048591 Supplier Thermo Scientific Acros Supplier No. 132690050
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 533-31-3 | C7H6O3 | 138.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-31-3
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00005827
InChI Key LUSZGTFNYDARNI-UHFFFAOYSA-N
Synonym sesamol, 3,4-methylenedioxy phenol, 5-hydroxy-1,3-benzodioxole, 3,4-methylenedioxyphenol, 2h-1,3-benzodioxol-5-ol, 3,4-methylendioxyphenol, methylene ether of oxyhydroquinone, phenol, 3,4-methylenedioxy, 5-benzodioxolol, unii-94iea0nv89
PubChem CID 68289
ChEBI CHEBI:9126
IUPAC Name 1,3-benzodioxol-5-ol
SMILES C1OC2=C(O1)C=C(C=C2)O

Specifications

Melting Point 63.0°C to 65.0°C
Color White to Beige
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 19, 66
Solubility Information Solubility in water: slightly soluble.
Formula Weight 138.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Sesamol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.