Learn More
Interscience™ Set of Tygon™ Tubings for Pump Head
Product Code. 12818291
Change view
Click to view available options
Diameter (Metric) Inner:
4.8 mm
6.4 mm
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
12818291 4.8 mm Each
17860087 6.4 mm Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12818291 Supplier Interscience™ Supplier No. 513011
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
20 cm long Tygon™ tubings for use with DiluFlow and FlexiPump pump heads.
  • Transparent
  • Flexible Tygon™ tubing
Specifications
Product Type Pump Tubing
Autoclavable Autoclavable
For Use With (Application) Adding Tubing to Dilutor Pumps
For Use With (Equipment) DiluFlow, DiluFlow Pro, DiluFlow Elite (5 kg), FlexiPump, FlexiPump Pro
Quantity 6 Tubings/Set
Diameter (Metric) Inner 4.8 mm